Assay Design
Gene Transcripts
- Gene Symbol
- Stmn1
- Entrez Gene ID
-
16765
- Gene Name
- stathmin 1
- Gene Aliases
-
19k, Lag, Lap18, Op18, P18, P19, Pig, Pp17, Pp18, Pp19, Pr22, Smn
- Chromosome Location
-
Chr.4: 134468320 - 134473843 on Build GRCm38
- Gene Symbol
- Gm11223
- Entrez Gene ID
-
100039888
- Gene Name
- stathmin 1 pseudogene
- Gene Aliases
-
OTTMUSG00000000267
- Chromosome Location
-
Chr.4: 69925201 - 69926170 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK030082.1 |
- |
- |
545 |
123 |
View Full Product Details