Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032400.2 | NP_115776.2 | 2 - 2 | 1306 | 155 |
GenBank mRNA | AF295367.1 | - | 2 - 2 | 1322 | 155 |
AK080866.1 | - | 2 - 2 | 1320 | 155 | |
BC096665.1 | - | 2 - 2 | 1279 | 155 |