Assay Design



Gene Transcripts

Gene Symbol
Wdr93
Entrez Gene ID
626359
Gene Name
WD repeat domain 93
Gene Aliases
EG626359
Chromosome Location
Chr.7: 79743135 - 79785952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037927.1 NP_001033016.1 8 - 9 888 120
GenBank mRNA AB205021.1 - 8 - 9 888 120
BC150920.1 - 7 - 8 858 120

View Full Product Details