Assay Design



Gene Transcripts

Gene Symbol
Filip1l
Entrez Gene ID
78749
Gene Name
filamin A interacting protein 1-like
Gene Aliases
4631422O05Rik, Doc1
Chromosome Location
Chr.16: 57353277 - 57572804 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040397.4 NP_001035487.2 2 - 3 438 96
GenBank mRNA AK146705.1 - 2 - 3 439 96
EU704258.1 - 2 - 3 285 96

View Full Product Details