Assay Design



Gene Transcripts

Gene Symbol
Krt36
Entrez Gene ID
16673
Gene Name
keratin 36
Gene Aliases
HRa-1, K36, Krt1-22, Krt1-5, MHRa-1
Chromosome Location
Chr.11: 100102012 - 100114634 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174099.1 NP_001167570.1 1 - 1 165 106
GenBank mRNA AK132500.1 - 1 - 1 181 106

View Full Product Details