Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025716.2 | NP_079992.1 | 1 - 2 | 88 | 135 |
GenBank mRNA | AK014623.1 | - | 1 - 2 | 93 | 135 |
AK080726.1 | - | 1 - 2 | 102 | 135 | |
AK082745.1 | - | 1 - 2 | 102 | 135 | |
BC014757.1 | - | 1 - 2 | 88 | 135 |