Assay Design
Gene Transcripts
- Gene Symbol
- Rhox4b
- Entrez Gene ID
-
57737
- Gene Name
- reproductive homeobox 4B
- Gene Aliases
-
5430432L21Rik, Ehox, Rhox4.2, Rhox4a
- Chromosome Location
-
Chr.X: 37432494 - 37437278 on Build GRCm38
- Gene Symbol
- Rhox4a
- Entrez Gene ID
-
664609
- Gene Name
- reproductive homeobox 4A
- Gene Aliases
-
Rhox4, Rhox4.1
- Chromosome Location
-
Chr.X: 37265318 - 37270136 on Build GRCm38
- Gene Symbol
- Rhox4c
- Entrez Gene ID
-
434759
- Gene Name
- reproductive homeobox 4C
- Gene Aliases
-
Rhox4.3, Rhox4b
- Chromosome Location
-
Chr.X: 37480337 - 37485124 on Build GRCm38
- Gene Symbol
- Rhox4d
- Entrez Gene ID
-
664610
- Gene Name
- reproductive homeobox 4D
- Gene Aliases
-
Rhox4.4, Rhox4c
- Chromosome Location
-
Chr.X: 37514389 - 37519176 on Build GRCm38
- Gene Symbol
- Rhox4e
- Entrez Gene ID
-
194856
- Gene Name
- reproductive homeobox 4E
- Gene Aliases
-
Rhox4, Rhox4.5, Rhox4d
- Chromosome Location
-
Chr.X: 37557412 - 37562282 on Build GRCm38
- Gene Symbol
- Rhox4f
- Entrez Gene ID
-
636177
- Gene Name
- reproductive homeobox 4F
- Gene Aliases
-
Rhox4.6, Rhox4e, Rhox4h
- Chromosome Location
-
Chr.X: 37602895 - 37607686 on Build GRCm38
- Gene Symbol
- Rhox4g
- Entrez Gene ID
-
664608
- Gene Name
- reproductive homeobox 4G
- Gene Aliases
-
Rhox4.6, Rhox4.7, Rhox4f
- Chromosome Location
-
Chr.X: 37646501 - 37651327 on Build GRCm38
View Full Product Details