Assay Design



Gene Transcripts

Gene Symbol
Bmp3
Entrez Gene ID
110075
Gene Name
bone morphogenetic protein 3
Gene Aliases
9130206H07, 9530029I04Rik
Chromosome Location
Chr.5: 98854432 - 98884052 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173404.3 NP_775580.1 2 - 2 1197 130
GenBank mRNA AK033644.1 - 2 - 2 835 130
AK035299.1 - 2 - 2 1223 130
AK035393.1 - 2 - 2 1186 130
AK037225.1 - 2 - 2 1178 130
AK040389.1 - 2 - 2 1198 130
BC117749.2 - 2 - 2 810 130

View Full Product Details