Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183300.2 | NP_899123.2 | 15 - 15 | 1948 | 121 |
GenBank mRNA | BC038255.1 | - | 4 - 4 | 443 | 121 |
BC046341.1 | - | 5 - 5 | 584 | 121 | |
BC058416.1 | - | 5 - 5 | 508 | 121 | |
BC079578.1 | - | 5 - 5 | 516 | 121 | |
DQ303413.1 | - | 15 - 15 | 1948 | 121 |