Assay Design
Gene Transcripts
- Gene Symbol
- Hist1h4a
- Entrez Gene ID
-
326619
- Gene Name
- histone cluster 1, H4a
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 23760795 - 23761249 on Build GRCm38
- Gene Symbol
- Hist1h4m
- Entrez Gene ID
-
100041230
- Gene Name
- histone cluster 1, H4m
- Gene Aliases
-
Gm11275, OTTMUSG00000000446
- Chromosome Location
-
Chr.13: 21811746 - 21812150 on Build GRCm38
- Gene Symbol
- Hist1h4n
- Entrez Gene ID
-
319161
- Gene Name
- histone cluster 1, H4n
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 21831793 - 21832158 on Build GRCm38
View Full Product Details