Assay Design



Gene Transcripts

Gene Symbol
Dgcr14
Entrez Gene ID
27886
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
AI462402, D16H22S1269E, Dgcr1, Dgsi, ES2, Es2el
Chromosome Location
Chr.16: 17900709 - 17911364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081633.1 NP_001075102.1 1 - 2 189 75
NM_022408.2 NP_071853.2 1 - 2 189 75
GenBank mRNA AF037256.1 - 1 - 2 188 75
AK088520.1 - 1 - 2 154 75
AK142684.1 - 1 - 2 154 75
AK148404.1 - 1 - 2 157 75
BC013711.1 - 1 - 2 168 75
BY069641.1 - 1 - 2 190 75

View Full Product Details