Assay Design



Gene Transcripts

Gene Symbol
Dgcr14
Entrez Gene ID
27886
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
AI462402, D16H22S1269E, Dgcr1, Dgsi, ES2, Es2el
Chromosome Location
Chr.16: 17900709 - 17911364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081633.1 NP_001075102.1 2 - 3 355 71
NM_022408.2 NP_071853.2 2 - 3 355 71
GenBank mRNA AF037256.1 - 2 - 3 354 71
AK088520.1 - 2 - 3 320 71
AK142684.1 - 2 - 3 320 71
AK148404.1 - 2 - 3 323 71
BC013711.1 - 2 - 3 334 71

View Full Product Details