Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130529.2 | NP_001124001.1 | 6 - 7 | 1358 | 80 |
NM_001291014.1 | NP_001277943.1 | 6 - 7 | 1112 | 80 | |
NM_172941.4 | NP_766529.3 | 6 - 7 | 1405 | 80 | |
GenBank mRNA | AK051711.1 | - | 6 - 7 | 694 | 80 |
AK139346.1 | - | 6 - 7 | 1648 | 80 | |
AK155196.1 | - | 6 - 7 | 935 | 80 | |
AK155490.1 | - | 6 - 7 | 987 | 80 | |
AK170924.1 | - | 6 - 7 | 935 | 80 | |
AK170989.1 | - | 6 - 7 | 940 | 80 | |
AK171190.1 | - | 6 - 7 | 987 | 80 | |
AY302242.1 | - | 5 - 6 | 886 | 80 | |
BC040205.1 | - | 5 - 6 | 890 | 80 |