Assay Design



Gene Transcripts

Gene Symbol
Mrgprb4
Entrez Gene ID
233230
Gene Name
MAS-related GPR, member B4
Gene Aliases
MrgB4
Chromosome Location
Chr.7: 48198213 - 48199178 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205795.1 NP_991364.1 1 - 1 41 68
GenBank mRNA BC107202.1 - 1 - 1 34 68
BC107203.1 - 1 - 1 34 68

View Full Product Details