Assay Design



Gene Transcripts

Gene Symbol
Kat6a
Entrez Gene ID
244349
Gene Name
K(lysine) acetyltransferase 6A
Gene Aliases
1500036M03, 9930021N24Rik, MOZ, Myst3, Zfp220
Chromosome Location
Chr.8: 22859516 - 22943262 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081149.1 NP_001074618.1 18 - 18 9024 111
GenBank mRNA AK148261.1 - 1 - 1 3352 111

View Full Product Details