Assay Design
Gene Transcripts
- Gene Symbol
- Phox2b
- Entrez Gene ID
-
18935
- Gene Name
- paired-like homeobox 2b
- Gene Aliases
-
Dilp1, NBPhox, Pmx2b
- Chromosome Location
-
Chr.5: 67094397 - 67099126 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_008888.3 |
NP_032914.1 |
3 - 3 |
2785 |
175 |
GenBank mRNA |
BC079610.1 |
- |
3 - 3 |
2786 |
175 |
View Full Product Details