Assay Design



Gene Transcripts

Gene Symbol
Sox11
Entrez Gene ID
20666
Gene Name
SRY (sex determining region Y)-box 11
Gene Aliases
1110038H03Rik, 6230403H02Rik, AI836553, end1
Chromosome Location
Chr.12: 27334268 - 27342718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009234.6 NP_033260.4 1 - 1 7440 87
GenBank mRNA AK004158.1 - 1 - 1 313 87
AK012306.1 - 1 - 1 1133 87

View Full Product Details