Assay Design



Gene Transcripts

Gene Symbol
Fgfr3
Entrez Gene ID
14184
Gene Name
fibroblast growth factor receptor 3
Gene Aliases
CD333, FR3, Fgfr-3, Flg-2, HBGFR, Mfr3, sam3
Chromosome Location
Chr.5: 33721724 - 33737068 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163215.2 NP_001156687.1 18 - 18 3673 128
NM_001163216.2 NP_001156688.1 16 - 16 3388 128
NM_001163217.2 NP_001156689.1 17 - 17 3448 128
NM_001205270.1 NP_001192199.1 18 - 18 3650 128
NM_008010.5 NP_032036.2 18 - 18 3647 128
GenBank mRNA AK143592.1 - 18 - 18 3583 128
AK163495.1 - 16 - 16 3226 128
BC053056.1 - 18 - 18 3557 128

View Full Product Details