Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009327.3 | NP_033353.2 | 10 - 10 | 2476 | 140 |
GenBank mRNA | AK141225.1 | - | 10 - 10 | 2282 | 140 |
BC080698.1 | - | 10 - 10 | 2478 | 140 | |
M57966.1 | - | - | 2456 | 140 |