Assay Design
Gene Transcripts
- Gene Symbol
- Hist1h2bm
- Entrez Gene ID
-
319186
- Gene Name
- histone cluster 1, H2bm
- Gene Aliases
-
H2b-291b
- Chromosome Location
-
Chr.13: 21722044 - 21722526 on Build GRCm38
- Gene Symbol
- Hist1h2ba
- Entrez Gene ID
-
319177
- Gene Name
- histone cluster 1, H2ba
- Gene Aliases
-
Th2b
- Chromosome Location
-
Chr.13: 23933725 - 23934156 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175663.2 |
NP_783594.1 |
1 - 1 |
307 |
85 |
GenBank mRNA |
BC126923.1 |
- |
1 - 1 |
307 |
85 |
- Gene Symbol
- Hist1h2bc
- Entrez Gene ID
-
68024
- Gene Name
- histone cluster 1, H2bc
- Gene Aliases
-
2610022J01Rik, H2bfs, R74621
- Chromosome Location
-
Chr.13: 23684199 - 23692480 on Build GRCm38
- Gene Symbol
- Hist1h2bf
- Entrez Gene ID
-
319180
- Gene Name
- histone cluster 1, H2bf
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 23573760 - 23574190 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178195.2 |
NP_835502.1 |
1 - 1 |
304 |
85 |
- Gene Symbol
- Hist1h2bg
- Entrez Gene ID
-
319181
- Gene Name
- histone cluster 1, H2bg
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 23571400 - 23571863 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178196.4 |
NP_835503.1 |
1 - 1 |
337 |
85 |
GenBank mRNA |
BC060304.1 |
- |
1 - 1 |
364 |
85 |
- Gene Symbol
- Hist1h2bh
- Entrez Gene ID
-
319182
- Gene Name
- histone cluster 1, H2bh
- Gene Aliases
-
H2b-221
- Chromosome Location
-
Chr.13: 23542923 - 23543444 on Build GRCm38
- Gene Symbol
- Hist1h2bj
- Entrez Gene ID
-
319183
- Gene Name
- histone cluster 1, H2bj
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 22043230 - 22043658 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178198.2 |
NP_835505.1 |
1 - 1 |
304 |
85 |
GenBank mRNA |
BC115792.1 |
- |
- |
296 |
85 |
- Gene Symbol
- Hist1h2bk
- Entrez Gene ID
-
319184
- Gene Name
- histone cluster 1, H2bk
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 22035821 - 22036320 on Build GRCm38
- Gene Symbol
- Hist1h2bl
- Entrez Gene ID
-
319185
- Gene Name
- histone cluster 1, H2bl
- Gene Aliases
-
H2b-291a
- Chromosome Location
-
Chr.13: 21715713 - 21716143 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178199.2 |
NP_835506.1 |
1 - 1 |
304 |
85 |
- Gene Symbol
- Hist1h2bp
- Entrez Gene ID
-
319188
- Gene Name
- histone cluster 1, H2bp
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 21787480 - 21801561 on Build GRCm38
- Gene Symbol
- Hist1h2bq
- Entrez Gene ID
-
665596
- Gene Name
- histone cluster 1, H2bq
- Gene Aliases
-
Gm13646, OTTMUSG00000013203
- Chromosome Location
-
Chr.13: 21806412 - 21810199 on Build GRCm38
- Gene Symbol
- Hist1h2br
- Entrez Gene ID
-
665622
- Gene Name
- histone cluster 1 H2br
- Gene Aliases
-
Gm11277, OTTMUSG00000000449
- Chromosome Location
-
Chr.13: 21833743 - 21837530 on Build GRCm38
View Full Product Details