Assay Design



Gene Transcripts

Gene Symbol
Psmg4
Entrez Gene ID
69666
Gene Name
proteasome (prosome, macropain) assembly chaperone 4
Gene Aliases
PAC-4, mPAC4
Chromosome Location
Chr.13: 34162964 - 34178172 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110515.2 NP_001103985.1 1 - 2 290 74
GenBank mRNA AK009887.1 - 1 - 2 175 74

View Full Product Details