Assay Design
Gene Transcripts
- Gene Symbol
- Cdh9
- Entrez Gene ID
-
12565
- Gene Name
- cadherin 9
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 16728549 - 16857097 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009869.1 |
NP_033999.1 |
4 - 5 |
807 |
77 |
View Full Product Details