Assay Design



Gene Transcripts

Gene Symbol
Sgcz
Entrez Gene ID
244431
Gene Name
sarcoglycan zeta
Gene Aliases
C230085N17Rik
Chromosome Location
Chr.8: 37517840 - 38661508 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145841.2 NP_665840.2 6 - 7 1366 65
GenBank mRNA AK041231.1 - 6 - 7 971 65
AK048960.1 - 6 - 7 1330 65
AK136779.1 - 6 - 7 1367 65
AY095374.1 - 5 - 6 582 65
AY250713.1 - 5 - 6 582 65
BC104048.1 - 5 - 6 582 65
BC104049.2 - 5 - 6 582 65

View Full Product Details