Assay Design



Gene Transcripts

Gene Symbol
Vmn2r2
Entrez Gene ID
100125586
Gene Name
vomeronasal 2, receptor 2
Gene Aliases
ENSMUSG00000043897
Chromosome Location
Chr.3: 64115730 - 64140657 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104592.1 NP_001098062.1 2 - 3 1143 139

View Full Product Details