Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
GenBank mRNA | U09772.1 | - | LIT1766 His1 |
1 - 2 | 150 | 95 |
U10269.1 | - | LIT1767 His1 |
1 - 2 | 150 | 95 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | zinc finger E-box binding homeobox 2, opposite strand | Zeb2os | Gm13476, OTTMUSG00000012358 | -163.0kb | Sense | Chr. - 45111138 - 45114087 on Build GRCm38 |
Downstream | RIKEN cDNA 1700019E08 gene | 1700019E08Rik | 417.4kb | Sense | Chr. - 45696605 - 45698447 on Build GRCm38 |