Assay Design
Gene Transcripts
- Gene Symbol
- Oas1b
- Entrez Gene ID
-
23961
- Gene Name
- 2'-5' oligoadenylate synthetase 1B
- Gene Aliases
-
Flv, L1, Mmu-L1, Oas1, Oias-2, Oias2, Wnv
- Chromosome Location
-
Chr.5: 120812635 - 120824163 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DT922876.1 |
- |
2 - 3 |
364 |
|
Chr. 5 - 120821222 - 120823557 [+] on Build GRCm38 |
97 |
GenBankmRNA |
BE288584.1 |
- |
1 - 2 |
65 |
|
Chr. 5 - 120822192 - 120823906 [+] on Build GRCm38 |
97 |
GenBankmRNA |
AI386448.1 |
- |
2 - 3 |
318 |
|
Chr. 5 - 120821259 - 120823579 [+] on Build GRCm38 |
97 |
GenBankmRNA |
BE369345.1 |
- |
1 - 2 |
97 |
|
Chr. 5 - 120822160 - 120823778 [+] on Build GRCm38 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
2'-5' oligoadenylate synthetase 1C |
Oas1c |
Oasl2, Oasl5 |
-9.6kb
|
Anti-sense |
Chr. - 120800194 - 120812540 on Build GRCm38 |
Downstream |
2'-5' oligoadenylate synthetase 1F |
Oas1f |
|
23.2kb
|
Sense |
Chr. - 120847367 - 120857986 on Build GRCm38 |
View Full Product Details