Assay Design
Gene Transcripts
- Gene Symbol
- Speer1
- Entrez Gene ID
-
70896
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 1
- Gene Aliases
-
4921519H07Rik, SPEER-1, Speer1-ps, Speer1-ps1
- Chromosome Location
-
Chr.5: 11340408 - 11346273 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001586.3 |
|
2 - 3 |
266 |
88 |
- Gene Symbol
- 4933402N22Rik
- Entrez Gene ID
-
545732
- Gene Name
- RIKEN cDNA 4933402N22 gene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 11918043 - 11922804 on Build GRCm38
- Gene Symbol
- Gm5861
- Entrez Gene ID
-
545728
- Gene Name
- predicted gene 5861
- Gene Aliases
-
EG545728
- Chromosome Location
-
Chr.5: 11182919 - 11187810 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC030042.1 |
- |
- |
251 |
88 |
- Gene Symbol
- Gm6455
- Entrez Gene ID
-
623849
- Gene Name
- predicted gene 6455
- Gene Aliases
-
EG623849
- Chromosome Location
-
Chr.5: 10865029 - 10870808 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003596.2 |
|
2 - 3 |
223 |
88 |
GenBank mRNA |
BC048648.1 |
- |
2 - 3 |
223 |
88 |
- Gene Symbol
- Gm6460
- Entrez Gene ID
-
623898
- Gene Name
- predicted gene 6460
- Gene Aliases
-
EG623898
- Chromosome Location
-
Chr.5: 11594956 - 11599480 on Build GRCm38
View Full Product Details