Assay Design



Gene Transcripts

Gene Symbol
Glyatl3
Entrez Gene ID
435528
Gene Name
glycine-N-acyltransferase-like 3
Gene Aliases
EG435528, Gm5683
Chromosome Location
Chr.17: 40904623 - 40920715 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145060.1 NP_001138532.1 3 - 4 314 101

View Full Product Details