Assay Design



Gene Transcripts

Gene Symbol
Atp6ap1l
Entrez Gene ID
435376
Gene Name
ATPase, H+ transporting, lysosomal accessory protein 1-like
Gene Aliases
EG435376
Chromosome Location
Chr.13: 90883439 - 90905355 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145879.1 NP_001139351.1 1 - 2 158 75
GenBank mRNA CX238982.1 - 1 - 2 99 75

View Full Product Details