Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012027.2 | NP_036157.2 | 10 - 11 | 1533 | 66 |
NM_201245.3 | NP_957697.1 | 10 - 11 | 1533 | 66 | |
GenBank mRNA | AK035435.1 | - | 4 - 5 | 823 | 66 |
AK044512.1 | - | 9 - 10 | 1175 | 66 | |
AK154807.1 | - | 11 - 12 | 1540 | 66 | |
AK155630.1 | - | 10 - 11 | 1314 | 66 | |
AK158296.1 | - | 10 - 11 | 1522 | 66 | |
AK170849.1 | - | 10 - 11 | 1517 | 66 | |
AK171089.1 | - | 10 - 11 | 1522 | 66 | |
BC039932.1 | - | 9 - 10 | 1057 | 66 | |
BC049803.1 | - | 10 - 11 | 1059 | 66 | |
BC063067.1 | - | 10 - 11 | 1496 | 66 | |
U73200.1 | - | 10 - 11 | 1393 | 66 |