Assay Design



Gene Transcripts

Gene Symbol
Mprip
Entrez Gene ID
26936
Gene Name
myosin phosphatase Rho interacting protein
Gene Aliases
9530046C02, AA536749, AI647711, C76423, RIP3, Rhoip3, mKIAA0864, p116Rip
Chromosome Location
Chr.11: 59662473 - 59780860 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012027.2 NP_036157.2 20 - 21 3122 86
NM_201245.3 NP_957697.1 20 - 21 3122 86
GenBank mRNA AB093269.1 - 8 - 9 4947 86
AK035435.1 - 14 - 15 2411 86
AK154807.1 - 21 - 22 3129 86
AK155630.1 - 20 - 21 2903 86
AK158296.1 - 20 - 21 3111 86
AK162530.1 - 18 - 19 2468 86
AK165710.1 - 5 - 6 1076 86
AK170849.1 - 20 - 21 3106 86
AK171089.1 - 20 - 21 3111 86
BC027417.1 - 6 - 7 1211 86
BC039932.1 - 19 - 20 2646 86
BC049803.1 - 20 - 21 2648 86
BC063067.1 - 20 - 21 3085 86
U73200.1 - 20 - 21 2982 86

View Full Product Details