Assay Design



Gene Transcripts

Gene Symbol
Strada
Entrez Gene ID
72149
Gene Name
STE20-related kinase adaptor alpha
Gene Aliases
2610019A05Rik, 6030402H20Rik, AI480680, E130112C09Rik
Chromosome Location
Chr.11: 106162974 - 106193604 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK021381.1 - 1 - 1 1151 91


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream LIM domain containing 2 Limd2 0610025L06Rik, AI413966 -19.5kb Sense Chr. - 106156256 - 106161039 on Build GRCm38
Downstream coiled-coil domain containing 47 Ccdc47 2610204L23Rik, C88307, asp4, calumin 17.6kb Sense Chr. - 106199354 - 106216367 on Build GRCm38

View Full Product Details