Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007223.3 | NP_001007224.1 | 3 - 3 | 2183 | 91 |
GenBank mRNA | AK029455.1 | - | 1 - 1 | 853 | 91 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | vesicle amine transport protein 1 homolog-like (T. californica) | Vat1l | 9430073I07, AI427515, mKIAA1576 | -52.5kb | Sense | Chr. - 114205582 - 114374074 on Build GRCm38 |
Downstream | WW domain-containing oxidoreductase | Wwox | 5330426P09Rik, 9030416C10Rik, WOX1 | 11.5kb | Sense | Chr. - 114439630 - 115352712 on Build GRCm38 |