Assay Design
Gene Transcripts
- Gene Symbol
- Pim1
- Entrez Gene ID
-
18712
- Gene Name
- proviral integration site 1
- Gene Aliases
-
Pim-1
- Chromosome Location
-
Chr.17: 29491045 - 29495465 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK047301.1 |
- |
- |
852 |
85 |
View Full Product Details