Assay Design
Gene Transcripts
- Gene Symbol
- Gprin2
- Entrez Gene ID
-
432839
- Gene Name
- G protein regulated inducer of neurite outgrowth 2
- Gene Aliases
-
C130040D06Rik, C230073P13, Gm286, mKIAA0514
- Chromosome Location
-
Chr.14: 34185688 - 34201692 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK048200.1 |
- |
1 - 1 |
942 |
95 |
AK082455.1 |
- |
1 - 1 |
205 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CX730911.1 |
- |
1 |
309 |
|
Chr. 14 - 34186136 - 34186770 [-] on Build GRCm38 |
95 |
GenBankmRNA |
BB651025.1 |
- |
1 |
203 |
|
Chr. 14 - 34185987 - 34186664 [-] on Build GRCm38 |
95 |
GenBankmRNA |
BB367967.2 |
- |
1 |
291 |
|
Chr. 14 - 34186104 - 34186741 [-] on Build GRCm38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
neuropeptide Y receptor Y4 |
Npy4r |
NYYR-D, Ppyr1, Y4 |
-33.7kb
|
Sense |
Chr. - 34145646 - 34152419 on Build GRCm38 |
Downstream |
synaptotagmin XV |
Syt15 |
CHR10SYT, E230025K04Rik, sytXV |
32.6kb
|
Anti-sense |
Chr. - 34220046 - 34230421 on Build GRCm38 |
View Full Product Details