Assay Design



Gene Transcripts

Gene Symbol
Nr1d1
Entrez Gene ID
217166
Gene Name
nuclear receptor subfamily 1, group D, member 1
Gene Aliases
A530070C09Rik, R75201
Chromosome Location
Chr.11: 98767932 - 98775377 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK053029.1 - 1 - 1 4352 72


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB525079.2 - 1 604 Chr. 11 - 98772754 - 98773399 [+] on Build GRCm38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream thyroid hormone receptor alpha Thra 6430529J03Rik, AW259572, Erba, Nr1a1, Rvr, T3R[a], T3Ralpha, Thra1, Thra2 -3.9kb Sense Chr. - 98741873 - 98765111 on Build GRCm38
Downstream male-specific lethal 1 homolog (Drosophila) Msl1 2810017F12Rik, 4121402D02Rik, 4930463F05Rik, AA682082, Msl-1 22.1kb Sense Chr. - 98795468 - 98807862 on Build GRCm38

View Full Product Details