Assay Design



Gene Transcripts

Gene Symbol
Mcu
Entrez Gene ID
215999
Gene Name
mitochondrial calcium uniporter
Gene Aliases
2010012O16Rik, AV064928, C10orf42, Ccdc109a, D130073L02Rik, Gm64
Chromosome Location
Chr.10: 59446984 - 59616692 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK166044.1 - 1 - 2 148 91


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY734336.1 - 1 - 2 148 Chr. 10 - 59534799 - 59616675 [-] on Build GRCm38 91


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream oncoprotein induced transcript 3 Oit3 EF-9, LZP -158.8kb Sense Chr. - 59422960 - 59441779 on Build GRCm38
Downstream mitochondrial calcium uptake 1 Micu1 C730016L05Rik, Calc, Cbara1 85.8kb Anti-sense Chr. - 59702482 - 59864134 on Build GRCm38

View Full Product Details