Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK177905.1 | - | 1 - 2 | 69 | 90 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | ras homolog gene family, member G | Rhog | 2810426G09Rik, Arhg, Sid10750 | -177.1kb | Anti-sense | Chr. - 102239123 - 102250118 on Build GRCm38 |
Downstream | ribonucleotide reductase M1 | Rrm1 | RnrM1 | 12.9kb | Sense | Chr. - 102441695 - 102469771 on Build GRCm38 |