Assay Design
Gene Transcripts
- Gene Symbol
- Ins2
- Entrez Gene ID
-
16334
- Gene Name
- insulin II
- Gene Aliases
-
AA986540, Ins-2, InsII, Mody, Mody4, proinsulin
- Chromosome Location
-
Chr.7: 142678656 - 142679708 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC066208.1 |
- |
2 - 3 |
229 |
72 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CF577777.1 |
- |
1 - 2 |
|
|
Chr. 7 - 142678656 - 142679685 [+] on Build GRCm38 |
72 |
GenBankmRNA |
BG881187.1 |
- |
2 - 3 |
228 |
|
Chr. 7 - 142678653 - 142679684 [-] on Build GRCm38 |
72 |
GenBankmRNA |
CF577776.1 |
- |
2 - 3 |
240 |
|
Chr. 7 - 142678656 - 142679685 [-] on Build GRCm38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
insulin-like growth factor 2, opposite strand |
Igf2os |
Igf2as, Peg8 |
-8.3kb
|
Anti-sense |
Chr. - 142659693 - 142670356 on Build GRCm38 |
Downstream |
tyrosine hydroxylase |
Th |
|
213.2kb
|
Sense |
Chr. - 142892776 - 142899966 on Build GRCm38 |
View Full Product Details