Assay Design
Gene Transcripts
- Gene Symbol
- Dpf2
- Entrez Gene ID
-
19708
- Gene Name
- D4, zinc and double PHD fingers family 2
- Gene Aliases
-
2210010M07Rik, BAF45D, Req, ubi-d4
- Chromosome Location
-
Chr.19: 5896516 - 5912871 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK019051.1 |
- |
1 - 2 |
77 |
72 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY708591.1 |
- |
1 - 2 |
75 |
|
Chr. 19 - 5911826 - 5912809 [-] on Build GRCm38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tigger transposable element derived 3 |
Tigd3 |
|
-16.7kb
|
Sense |
Chr. - 5891138 - 5895166 on Build GRCm38 |
Downstream |
predicted gene, 42067 |
Gm42067 |
|
159.0
|
Anti-sense |
Chr. - 5912968 - 5914831 on Build GRCm38 |
View Full Product Details