Assay Design
Gene Transcripts
- Gene Symbol
- 5330430B06Rik
- Entrez Gene ID
-
78280
- Gene Name
- RIKEN cDNA 5330430B06 gene
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK019920.1 |
- |
1 - 1 |
649 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
phosphodiesterase 10A |
Pde10a |
|
|
Sense |
Chr. - 8525187 - 8986648 on Build GRCm38 |
Downstream |
RIKEN cDNA 1700010I14 gene |
1700010I14Rik |
|
375.4kb
|
Sense |
Chr. - 8988320 - 9008319 on Build GRCm38 |
View Full Product Details