Assay Design
Gene Transcripts
- Gene Symbol
- Tmeff2
- Entrez Gene ID
-
56363
- Gene Name
- transmembrane protein with EGF-like and two follistatin-like domains 2
- Gene Aliases
-
4832418D20Rik, 7630402F16Rik, TR-2
- Chromosome Location
-
Chr.1: 50909962 - 51187270 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK020188.1 |
- |
- |
134 |
76 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY720399.1 |
- |
1 - 2 |
134 |
|
Chr. 1 - 50900647 - 50913324 [+] on Build GRCm38 |
76 |
GenBankmRNA |
BY724413.1 |
- |
1 - 2 |
134 |
|
Chr. 1 - 50900647 - 50914650 [+] on Build GRCm38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
predicted gene 28321 |
Gm28321 |
|
-82.9kb
|
Sense |
Chr. - 50812475 - 50817701 on Build GRCm38 |
Downstream |
RIKEN cDNA 9330175M20 gene |
9330175M20Rik |
|
305.5kb
|
Anti-sense |
Chr. - 51218386 - 51333779 on Build GRCm38 |
View Full Product Details