Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK140616.1 | - | 1 - 2 | 1225 | 73 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase) | Alg1 | HMAT1, HMT1 | -9.9kb | Sense | Chr. - 5233621 - 5244907 on Build GRCm38 |
Downstream | RNA binding protein, fox-1 homolog (C. elegans) 1 | Rbfox1 | A2bp, A2bp1, Hrnbp1, fox-1 | 595.2kb | Sense | Chr. - 5884793 - 7412480 on Build GRCm38 |