Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE946802.1 |
- |
1 |
|
|
Chr. 11 - 62515720 - 62516040 [-] on Build GRCm38 |
77 |
GenBankmRNA |
BM219285.2 |
- |
1 |
|
|
Chr. 11 - 62514846 - 62516035 [-] on Build GRCm38 |
77 |
GenBankmRNA |
EH096498.1 |
- |
|
|
|
Chr. - - [-] on Build |
77 |
GenBankmRNA |
BF456177.1 |
- |
1 |
|
|
Chr. 11 - 62515720 - 62515960 [-] on Build GRCm38 |
77 |
GenBankmRNA |
AK077687.1 |
- |
|
1555 |
|
Chr. - - [-] on Build |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
phosphatidylinositol glycan anchor biosynthesis, class L |
Pigl |
Gm737 |
-1.8kb
|
Anti-sense |
Chr. - 62458460 - 62513900 on Build GRCm38 |
Downstream |
centromere protein V |
Cenpv |
3110013H01Rik, AA671303, Prr6 |
9.0kb
|
Sense |
Chr. - 62524944 - 62539502 on Build GRCm38 |
View Full Product Details