Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113558.2 | NP_001107030.1 | 4 - 5 | 452 | 56 |
NM_026754.3 | NP_081030.1 | 3 - 4 | 386 | 56 | |
GenBank mRNA | AK003750.1 | - | 3 - 4 | 328 | 56 |
BC115608.2 | - | 4 - 5 | 355 | 56 | |
BC115609.2 | - | 4 - 5 | 355 | 56 | |
EF529510.1 | - | 4 - 5 | 423 | 56 | |
EU368184.1 | - | 4 - 5 | 452 | 56 | |
FJ217399.1 | - | 4 - 5 | 390 | 56 | |
FJ217400.1 | - | 3 - 4 | 324 | 56 |