Assay Design



Gene Transcripts

Gene Symbol
Gm17660
Entrez Gene ID
100271704
Gene Name
predicted gene, 17660
Gene Aliases
Scpppq1
Chromosome Location
Chr.5: 104070060 - 104077608 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163772.1 NP_001157244.1 6 - 7 287 113
GenBank mRNA EU642618.1 - 6 - 7 287 113
HM015622.1 - 6 - 7 315 113
HM015623.1 - 6 - 7 271 113

View Full Product Details