Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146342.2 | NP_001139814.2 | 2 - 3 | 503 | 80 |
NM_001167818.1 | NP_001161290.1 | 4 - 5 | 752 | 80 | |
GenBank mRNA | AK170321.1 | - | 2 - 3 | 504 | 80 |
BC052355.1 | - | 4 - 5 | 583 | 80 | |
DQ788834.1 | - | 4 - 5 | 529 | 80 |