Assay Design



Gene Transcripts

Gene Symbol
Gm17365
Entrez Gene ID
100312956
Gene Name
predicted gene, 17365
Gene Aliases
Pate-dj, Pate3
Chromosome Location
Chr.9: 35645113 - 35648202 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167592.2 NP_001161064.1 2 - 3 261 99
GenBank mRNA EU004071.1 - 2 - 3 261 99
EU703626.1 - 2 - 3 178 99

View Full Product Details