Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024190.2 | NP_077152.1 | 1 - 1 | 450 | 72 |
GenBank mRNA | AK012996.1 | - | 1 - 1 | 400 | 72 |
AK014091.1 | - | 1 - 1 | 417 | 72 | |
AK151071.1 | - | 1 - 1 | 419 | 72 | |
AK167614.1 | - | 1 - 1 | 388 | 72 | |
BC002229.1 | - | 1 - 1 | 386 | 72 | |
BC034442.1 | - | 1 - 1 | 374 | 72 | |
CT010307.1 | - | 1 - 1 | 308 | 72 |