Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168470.1 | NP_001161942.1 | 2 - 2 | 448 | 96 |
NM_025318.3 | NP_079594.1 | 2 - 2 | 399 | 96 | |
GenBank mRNA | AK002385.1 | - | 2 - 2 | 384 | 96 |
AK003198.1 | - | 2 - 2 | 394 | 96 | |
AK004149.1 | - | 2 - 2 | 395 | 96 | |
AK012769.1 | - | 2 - 2 | 384 | 96 | |
BC022104.1 | - | 2 - 2 | 370 | 96 |